phynomic lite Safety Gloves, 60040 Series
: UVEX
The phynomic series is ideal for all types of work where a high level of feeling is required. These safety gloves are like a 'second skin' and are especially light and flexible.
- Exceptionally light and flexible
- Good grip in dry and slightly damp areas
- Food-safe
- Excellent skin tolerance and free from pollutants
The uvex phynomic lite is extremely lightweight, with a composition that makes it suited to particularly delicate tasks. They are free of accelerators and dermatologically tested for skin compatibility, making them ideal for allergy sufferers. These gloves also offer a very high level of breathability with the porous coating, which reduces sweating.
- Catalog No:
- UVEX6004005
- UVEX6004012
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- Glove sizeColour
- : 6004005: UVEX6004005
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018 - : 6004012: UVEX6004012
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018