Constant climatic chambers with Peltier, HPP series
Supplier: Memmert
MEMMHPP400EA
14660
GBP
MEMMHPP400
Constant climatic chambers with Peltier, HPP series
The HPP range of climatic chambers offers precise temperature control from 0 up to +70 °C with active humidification and dehumidification from 10 to 90% RH. All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. Units have Peltier technology for energy efficient cooling and heating. The Peltier elements can be controlled individually ensuring homogenous temperature and humidity distribution in the chamber. All models feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances, and can send an automatic alarm message to one or several email addresses. Complex ramping processes can be programmed quickly and easily. Chambers are ideal for stability tests and storage in controlled environment and conditioning.
- Intuitive, user friendly TwinDISPLAY with two TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), program time, relative humidity, illumination, time zones and daylight saving time
- Two PT100 sensors in a four wire circuit for mutual monitoring, taking over functions in case of an error
- The HeatBALANCE function allows application-specific adjustment of heating power distribution (balance) between the upper and lower heating / cooling groups (Peltier elements)
- Logged protocol data is displayed on the ControlCOCKPIT (max. 10000 values correspond to approx. 1 week)
- Ethernet interface for reading out the protocol log, for uploading and implementing programs and for online logging
- Multiple over-temperature protection with audible and visual alarm selectable on display: Electronic temperature monitoring TWW (independent adjustable temperature safety device Class 3.3) or TWB adjustable temperature limiter safety device Class 2 which protect the samples and the chamber. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual min./max. values for over- or under-temperature
Ordering information: Supplied without grids, please order required stainless steel grids, perforated shelves or reinforced grids separately. Range of accessories and factory fitted options such as LED light modules, access ports and interior sockets are available on request. Please contact VWR for further details prior to ordering, as these cannot be retro-fitted.
Caution: For details of the range of shelves and grids, please visit vwr.com or contact your local VWR sales office.
New Product
Limited Time Sale
Select items below are currently on sale
Services: Memmert HPP climatic chambers have a 3 year warranty period as standard. This is an additional 1 year extension to that 3 year period, giving 4 years in total.
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart