Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.
Primaide™ system manager
Supplier: VWR Collection
Primaide™
903-0743EA 0 GBP InStock
903-0743 903-0708
Primaide™ system manager
Software Laboratory Software
Chromatography Data System (CDS) for control of the VWR Hitachi Primaide HPLC Systems.
Functions for: Method creation, data acquisition, chromatograph data processing, DAD data processing and report generation.
PC requirements:
OS: Windows 7 32/64 bit or Windows 10 64 bit
CPU: Intel® Core2 Duo 2 GHz or higher
RAM: 2 GB or more required
Hard disk: At least 2 GB of free space on a partition containing the Windows® 7 system files
Display: Capable of supporting 1280x1024 pixels (256 colors or higher)
Caution: Not 21 CFR Part 11 Compliant, therefore not suitable for GMP/GLP environments. If 21 CFR Part 11 compliant software is required please check DataApex ClarityVA or Agilent Openlab in our webshop.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty