MF-Millipore™ Membrane Filters
Supplier: Merck
Certificates
MF-Millipore™ filters without Triton® surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Biologically inert mixtures of cellulose acetate and cellulose nitrate.
- Catalog No:
- 512-0277
- MILFRAWP03700
- MILFRAWG04700
- 512-0240
- MILFSSWP01300
- MILFHABP04700
- MILFHABP02500
- MILFGSWP03700
- MILFAABPC5000
- MILFRAWG02500
- MILFRAWP01300
- MILFVSWP14250
- 512-0283
- 512-0249
- MILFAAWP01300
- MILFHAWG01300
- MILFHATF14250
- MILFHATF10050
- 512-0256
- MILFHATF09025
- MILFVSWP09025
- MILFAAWG04700
- MILFHAWP09000
- 512-0257
- MILFHAWP05000
- MILFAAWG03700
- MILFVSWP01300
- MILFAAWG0250C
- MILFAAWG02500
- MILFGSTF04700
- MILFHATF02500
- MILFRAWP14250
- MILFHATF01300
- MILFHAWP03700
- 512-0258
- MILFVCWP04700
- MILFPHWP04700
- MILFDAWP04700
- MILFGSWP09000
- MILFAABP04700
- MILFDAWP01300
- MILFAAWG01300
- MILFGSTF09000
- MILFAABP02500
- MILFHAWP0470M
- MILFGSTF02500
- MILFHAWP02400
- MILFHAWG05000
- MILFGSWP14250
- MILFHATF04700
- MILFSMWP03700
- MILFHATF08250
- MILFSMWP09025
- MILFSMWP14250
- MILFAAWP14250
- MILFRATF14250
- MILFRAWG0250C
- MILFGSTF01300
- MILFAAWP09000
- MILFHAWP04700F1
- MILFAABG04700
- MILFRAWP05000
- MILFAABG03700
- MILFHAWG04705
- MILFHAWG04700
- MILFAABG02500
- MILFHAWG03700
- MILFAABG01300
- MILFHAWG02500
- 512-0273
- 512-0250
- 512-0275
- MILFSSWP14250
- MILFHAWP14250
- MILFSCWP0190R
- 512-0276
- 512-0251
- MILFVMWP01300
- MILFSSWP09025
- MILFVMWP02500
- 512-0281
- MILFRAWP02500
- MILFVMWP09025
- MILFAAWP0470M
- MILFAAWP037PM
- MILFAAWP037P0
- MILFHAEP04700
- MILFHABG04700
- MILFHABG02500
- MILFGSTF14250
- MILFHABG01300
- MILFPHWP02500
- MILFPHWP09025
- MILFPHWP14250
- MILFSCWP01300
- MILFRAWP09025
- 512-0270
- MILFSMWP0190R
- MILFVCWP01300
- 512-0274
- MILFDAWP14250
- MILFHATF08550
- MILFDAWP09025
- 512-0267
- MILFAAWP03700
- 512-0241
- 512-0271
- MILFSCWP14250
- MILFDAWP02500
- MILFVCWP02500
- MILFSCWP09025
- 512-0282
- MILFVCWP14250
- MILFVCWP09025
- Type:
- SSWP, white, plain
- RAWP, white, plain
- RAWG, white, gridded
- AAWP, white, plain
- SSWP, white, plain
- HABP, black, plain
- HABP, black, plain
- GSWP, white, plain, with thin pad
- AABP, black, plain
- RAWG, white, gridded
- RAWP, white, plain
- VSWP, white, plain
- VSWP, white, plain
- GSWP, white, plain
- AAWP, white, plain
- HAWG, white, gridded
- HATF, white, plain, Triton®-free
- HATF, white, plain, Triton®-free
- HAWP, white, plain
- HATF, white, plain, Triton®-free
- VSWP, white, plain
- AAWG, white, gridded
- HAWP, white, plain
- HAWP, white, plain
- HAWP, white, plain
- AAWG, white, gridded, with thin pad
- VSWP, white, plain
- AAWG, white, gridded
- AAWG
- GSTF, white, plain, Triton®-free
- HATF, white, plain, Triton®-free
- RAWP, white, plain
- HATF, white, plain, Triton®-free
- HAWP, white, plain, with thin pad
- HAWP, white, plain
- VCWP, white, plain
- PHWP, white, plain
- DAWP, white, plain
- GSWP, white, plain
- AABP, black, plain
- DAWP, white, plain
- AAWG, white, gridded
- GSTF, white, plain, Triton®-free
- AABP, black, plain
- HAWP, white, plain
- GSTF, white, plain, Triton®-free
- HAWP, white, plain
- HAWG, white, gridded
- GSWP, white, plain
- HATF, white, plain, Triton®-free
- SMWP, white, plain, with thin pad
- HATF, white, plain, Triton®-free
- SMWP, white, plain
- SMWP, white, plain
- AAWP, white, plain
- RATF, white, plain, Triton®-free
- RAWG, white, gridded
- GSTF, white, plain, Triton®-free
- AAWP, white, plain
- HAWP, white, plain
- AABG, black, gridded
- RAWP, white, plain
- AABG, black, gridded, with thin pad
- HAWG, white, gridded
- HAWG, white, gridded
- AABG, black, gridded
- HAWG, white, gridded, with thin pad
- AABG, black, gridded
- HAWG, white, gridded
- SMWP, white, plain
- GSWP, white, plain
- SMWP, white, plain
- SSWP, white, plain
- HAWP, white, plain
- SCWP, white, plain
- SSWP, white, plain
- GSWP, white, plain
- VMWP, white, plain
- SSWP, white, plain
- VMWP, white, plain
- VMWP, white, plain
- RAWP, white, plain
- VMWP, white, plain
- AAWP, white, plain
- AAWP, white, plain, with thick pad
- AAWP, white, plain, with thick pad
- White, plain
- HABG, black, gridded
- HABG, black, gridded
- GSTF, white, plain, Triton®-free
- HABG, black, gridded
- PHWP, white, plain
- PHWP, white, plain
- PHWP, white, plain
- SCWP, white, plain
- RAWP, white, plain
- SCWP, white, plain
- SMWP, white, plain
- VCWP, white, plain
- SMWP, white, plain
- DAWP, white, plain
- HATF, white, plain, Triton®-free
- DAWP, white, plain
- RAWP, white, plain
- AAWP, white, plain, with thin pad
- AAWP, white, plain
- SCWP, white, plain
- SCWP, white, plain
- DAWP, white, plain
- VCWP, white, plain
- SCWP, white, plain
- VSWP, white, plain
- VCWP, white, plain
- VCWP, white, plain
- Pore size:
- 3,00 µm
- 1,20 µm
- 1,20 µm
- 0,80 µm
- 3,00 µm
- 0,45 µm
- 0,45 µm
- 0,22 µm
- 0,8 µm
- 1,20 µm
- 1,20 µm
- 0,025 µm
- 0,025 µm
- 0,22 µm
- 0,80 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,025 µm
- 0,80 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,80 µm
- 0,025 µm
- 0,80 µm
- 0,80 µm
- 0,22 µm
- 0,45 µm
- 1,20 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,10 µm
- 0,30 µm
- 0,65 µm
- 0,22 µm
- 0,80 µm
- 0,65 µm
- 0,80 µm
- 0,22 µm
- 0,80 µm
- 0,45 µm
- 0,22 µm
- 0,45 µm
- 0,45 µm
- 0,22 µm
- 0,45 µm
- 5,00 µm
- 0,45 µm
- 5,00 µm
- 5,00 µm
- 0,80 µm
- 1,20 µm
- 1,20 µm
- 0,22 µm
- 0,80 µm
- 0,45 µm
- 0,80 µm
- 1,20 µm
- 0,80 µm
- 0,45 µm
- 0,45 µm
- 0,80 µm
- 0,45 µm
- 0,80 µm
- 0,45 µm
- 5,00 µm
- 0,22 µm
- 5,00 µm
- 3,00 µm
- 0,45 µm
- 8,00 µm
- 3,00 µm
- 0,22 µm
- 0,05 µm
- 3,00 µm
- 0,05 µm
- 0,05 µm
- 1,20 µm
- 0,05 µm
- 0,80 µm
- 0,80 µm
- 0,80 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,22 µm
- 0,45 µm
- 0,30 µm
- 0,30 µm
- 0,30 µm
- 8,00 µm
- 1,20 µm
- 8,00 µm
- 5,00 µm
- 0,10 µm
- 5,00 µm
- 0,65 µm
- 0,45 µm
- 0,65 µm
- 1,20 µm
- 0,80 µm
- 0,80 µm
- 8,00 µm
- 8,00 µm
- 0,65 µm
- 0,10 µm
- 8,00 µm
- 0,025 µm
- 0,10 µm
- 0,10 µm
- Thickness:
- 180 µm
- 170 µm
- 170 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- —
- 170 µm
- 170 µm
- 105 µm
- 105 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 105 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 105 µm
- 180 µm
- 150 µm
- 180 µm
- 180 µm
- 170 µm
- 180 µm
- 180 µm
- 180 µm
- 105 µm
- 170 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- -
- 180 µm
- 180 µm
- 160 µm
- 180 µm
- 160 µm
- 160 µm
- 180 µm
- 180 µm
- 170 µm
- 180 µm
- 180 µm
- 150 µm
- 180 µm
- 150 µm
- 180 µm
- 150 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 160 µm
- 180 µm
- 160 µm
- 180 µm
- 180 µm
- 160 µm
- 180 µm
- 180 µm
- 105 µm
- 180 µm
- 105 µm
- 105 µm
- 170 µm
- 105 µm
- 180 µm
- 180 µm
- 180 µm
- 150 µm
- 180 µm
- 180 µm
- 180 µm
- 180 µm
- 170 µm
- 170 µm
- 170 µm
- 160 µm
- 170 µm
- 160 µm
- 160 µm
- 105 µm
- 160 µm
- 180 µm
- 180 µm
- 180 µm
- 170 µm
- 180 µm
- 180 µm
- 160 µm
- 160 µm
- 180 µm
- 105 µm
- 160 µm
- 105 µm
- 105 µm
- 105 µm
- Water flow rate:
- 320 ml/min/cm²
- 270 ml/min/cm²
- 270 ml/min/cm²
- 190 ml/min/cm²
- 320 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 18 ml/min/cm²
- —
- 270 ml/min/cm²
- 270 ml/min/cm²
- 0,15 ml/min/cm²
- 0,15 ml/min/cm²
- 18 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 0,15 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 190 ml/min/cm²
- 0,15 ml/min/cm²
- 190 ml/min/cm²
- 190 ml/min/cm²
- 18 ml/min/cm²
- 270 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 1,5 ml/min/cm²
- 32 ml/min/cm²
- 140 ml/min/cm²
- 18 ml/min/cm²
- 190 ml/min/cm²
- 140 ml/min/cm²
- 190 ml/min/cm²
- 18 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 18 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 18 ml/min/cm²
- 580 ml/min/cm²
- 580 ml/min/cm²
- 580 ml/min/cm²
- 190 ml/min/cm²
- 270 ml/min/cm²
- 270 ml/min/cm²
- 18 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 190 ml/min/cm²
- 270 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 580 ml/min/cm²
- 18 ml/min/cm²
- 580 ml/min/cm²
- 320 ml/min/cm²
- 60 ml/min/cm²
- 620 ml/min/cm²
- 320 ml/min/cm²
- 18 ml/min/cm²
- 0,74 ml/min/cm²
- 320 ml/min/cm²
- 0,74 ml/min/cm²
- 0,74 ml/min/cm²
- 270 ml/min/cm²
- 0,74 ml/min/cm²
- 190 ml/min/cm²
- 190 ml/min/cm²
- 190 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 60 ml/min/cm²
- 18 ml/min/cm²
- 60 ml/min/cm²
- 32 ml/min/cm²
- 32 ml/min/cm²
- 32 ml/min/cm²
- 620 ml/min/cm²
- 270 ml/min/cm²
- 620 ml/min/cm²
- 580 ml/min/cm²
- 1,5 ml/min/cm²
- 580 ml/min/cm²
- 140 ml/min/cm²
- 140 ml/min/cm²
- 270 ml/min/cm²
- 190 ml/min/cm²
- 190 ml/min/cm²
- 620 ml/min/cm²
- 620 ml/min/cm²
- 140 ml/min/cm²
- 1,5 ml/min/cm²
- 620 ml/min/cm²
- 0,15 ml/min/cm²
- 1,5 ml/min/cm²
- 1,5 ml/min/cm²
- Airflow rate:
- 30 L/min/cm²
- 20 L/min/cm²
- 20 L/min/cm²
- 16 L/min/cm²
- 30 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 2 L/min/cm²
- —
- 20 L/min/cm²
- 20 L/min/cm²
- 0,15 L/min/cm²
- 0,15 L/min/cm²
- 2 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 0,15 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 16 L/min/cm²
- 0,15 L/min/cm²
- 16 L/min/cm²
- 16 L/min/cm²
- 2 L/min/cm²
- 20 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 0,4 L/min/cm²
- 3 L/min/cm²
- 9 L/min/cm²
- 2 L/min/cm²
- 16 L/min/cm²
- 9 L/min/cm²
- 16 L/min/cm²
- 2 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 2 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 2 L/min/cm²
- 32 L/min/cm²
- 32 L/min/cm²
- 32 L/min/cm²
- 16 L/min/cm²
- 20 L/min/cm²
- 20 L/min/cm²
- 2 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 16 L/min/cm²
- 20 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 32 L/min/cm²
- 2 L/min/cm²
- 32 L/min/cm²
- 30 L/min/cm²
- 4 L/min/cm²
- 65 L/min/cm²
- 30 L/min/cm²
- 2 L/min/cm²
- 0,25 L/min/cm²
- 30 L/min/cm²
- 0,25 L/min/cm²
- 0,25 L/min/cm²
- 20 L/min/cm²
- 0,25 L/min/cm²
- 16 L/min/cm²
- 16 L/min/cm²
- 16 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 4 L/min/cm²
- 2 L/min/cm²
- 4 L/min/cm²
- 3 L/min/cm²
- 3 L/min/cm²
- 3 L/min/cm²
- 65 L/min/cm²
- 20 L/min/cm²
- 65 L/min/cm²
- 32 L/min/cm²
- 0,4 L/min/cm²
- 32 L/min/cm²
- 9 L/min/cm²
- 9 L/min/cm²
- 20 L/min/cm²
- 16 L/min/cm²
- 16 L/min/cm²
- 65 L/min/cm²
- 65 L/min/cm²
- 9 L/min/cm²
- 0,4 L/min/cm²
- 65 L/min/cm²
- 0,15 L/min/cm²
- 0,4 L/min/cm²
- 0,4 L/min/cm²
- Max. temperature:
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- —
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
- 75 °C
Standard MF-Millipore™ membranes, white, plain
Product Information
- Catalog #Supplier #TypePore sizeØPkAvailabilityPrice
- 512-0240AAWP02500AAWP, white, plain0,80 µm25 mm1001 * 100 items£115.00: AAWP02500: MILFAAWP02500
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFGSWP03700GSWP03700GSWP, white, plain, with thin pad0,22 µm37 mm1001 * 100 items£170.00: GSWP03700: MILFGSWP03700
Specifications
TypeGSWP, white, plain, with thin padEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFRAWP03700RAWP03700RAWP, white, plain1,20 µm37 mm1001 * 100 items£161.00: RAWP03700: MILFRAWP03700
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFSSWP01300SSWP01300SSWP, white, plain3,00 µm13 mm1001 * 100 items£99.90: SSWP01300: MILFSSWP01300
Specifications
TypeSSWP, white, plainEnvironmentally PreferablePore size3,00 µmThickness180 µmWater flow rate320 ml/min/cm²Airflow rate30 L/min/cm²Max. temperature75 °C - 512-0277SSWP04700SSWP, white, plain3,00 µm47 mm1001 * 100 items£165.00: SSWP04700: MILFSSWP04700
Specifications
TypeSSWP, white, plainEnvironmentally PreferablePore size3,00 µmThickness180 µmWater flow rate320 ml/min/cm²Airflow rate30 L/min/cm²Max. temperature75 °C - MILFAAWP01300AAWP01300AAWP, white, plain0,80 µm13 mm1001 * 100 items£99.90: AAWP01300: MILFAAWP01300
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWP14250AAWP14250AAWP, white, plain0,80 µm142 mm501 * 50 items£371.00: AAWP14250: MILFAAWP14250
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFDAWP01300DAWP01300DAWP, white, plain0,65 µm13 mm1001 * 100 items£99.90: DAWP01300: MILFDAWP01300
Specifications
TypeDAWP, white, plainEnvironmentally PreferablePore size0,65 µmThickness180 µmWater flow rate140 ml/min/cm²Airflow rate9 L/min/cm²Max. temperature75 °C - MILFDAWP04700DAWP04700DAWP, white, plain0,65 µm47 mm1001 * 100 items£159.00: DAWP04700: MILFDAWP04700
Specifications
TypeDAWP, white, plainEnvironmentally PreferablePore size0,65 µmThickness180 µmWater flow rate140 ml/min/cm²Airflow rate9 L/min/cm²Max. temperature75 °C - 512-0249GSWP01300GSWP, white, plain0,22 µm13 mm1001 * 100 items£101.00: GSWP01300: MILFGSWP01300
Specifications
TypeGSWP, white, plainEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFGSWP09000GSWP09000GSWP, white, plain0,22 µm90 mm1001 * 100 items£407.00: GSWP09000: MILFGSWP09000
Specifications
TypeGSWP, white, plainEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFGSWP14250GSWP14250GSWP, white, plain0,22 µm142 mm501 * 50 items£378.00: GSWP14250: MILFGSWP14250
Specifications
TypeGSWP, white, plainEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - 512-0256HAWP01300HAWP, white, plain0,45 µm13 mm1001 * 100 items£96.70: HAWP01300: MILFHAWP01300
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP02400HAWP02400HAWP, white, plain0,45 µm24 mm1001 * 100 items£99.90: HAWP02400: MILFHAWP02400
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - 512-0258HAWP04700HAWP, white, plain0,45 µm47 mm1001 * 100 items£169.00: HAWP04700: MILFHAWP04700
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP0470MHAWP0470MHAWP, white, plain0,45 µm47 mm501 * 50 items£625.00: HAWP0470M: MILFHAWP0470M
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP03700HAWP03700HAWP, white, plain, with thin pad0,45 µm37 mm1001 * 100 items£179.00: HAWP03700: MILFHAWP03700
Specifications
TypeHAWP, white, plain, with thin padEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFPHWP04700PHWP04700PHWP, white, plain0,30 µm47 mm1001 * 100 items£313.00: PHWP04700: MILFPHWP04700
Specifications
TypePHWP, white, plainEnvironmentally PreferablePore size0,30 µmThickness170 µmWater flow rate32 ml/min/cm²Airflow rate3 L/min/cm²Max. temperature75 °C - MILFRAWP01300RAWP01300RAWP, white, plain1,20 µm13 mm1001 * 100 items£96.70: RAWP01300: MILFRAWP01300
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFRAWP14250RAWP14250RAWP, white, plain1,20 µm142 mm501 * 50 items£370.00: RAWP14250: MILFRAWP14250
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFSMWP09025SMWP09025SMWP, white, plain5,00 µm90 mm251 * 25 items£142.00: SMWP09025: MILFSMWP09025
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - MILFSMWP14250SMWP14250SMWP, white, plain5,00 µm142 mm501 * 50 items£402.00: SMWP14250: MILFSMWP14250
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - MILFSMWP03700SMWP03700SMWP, white, plain, with thin pad5,00 µm37 mm1001 * 100 items£183.00: SMWP03700: MILFSMWP03700
Specifications
TypeSMWP, white, plain, with thin padEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - MILFVCWP04700VCWP04700VCWP, white, plain0,10 µm47 mm1001 * 100 items£365.00: VCWP04700: MILFVCWP04700
Specifications
TypeVCWP, white, plainEnvironmentally PreferablePore size0,10 µmThickness105 µmWater flow rate1,5 ml/min/cm²Airflow rate0,4 L/min/cm²Max. temperature75 °C - 512-0283VSWP04700VSWP, white, plain0,025 µm47 mm1001 * 100 items£353.00: VSWP04700: MILFVSWP04700
Specifications
TypeVSWP, white, plainEnvironmentally PreferablePore size0,025 µmThickness105 µmWater flow rate0,15 ml/min/cm²Airflow rate0,15 L/min/cm²Max. temperature75 °C - MILFVSWP09025VSWP09025VSWP, white, plain0,025 µm90 mm251 * 25 items£307.00: VSWP09025: MILFVSWP09025
Specifications
TypeVSWP, white, plainEnvironmentally PreferablePore size0,025 µmThickness105 µmWater flow rate0,15 ml/min/cm²Airflow rate0,15 L/min/cm²Max. temperature75 °C - MILFVSWP14250VSWP14250VSWP, white, plain0,025 µm142 mm501 * 50 items£595.00: VSWP14250: MILFVSWP14250
Specifications
TypeVSWP, white, plainEnvironmentally PreferablePore size0,025 µmThickness105 µmWater flow rate0,15 ml/min/cm²Airflow rate0,15 L/min/cm²Max. temperature75 °C - 512-0257HAWP02500HAWP, white, plain0,45 µm25 mm1001 * 100 items£113.00: HAWP02500: MILFHAWP02500
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP05000HAWP05000HAWP, white, plain0,45 µm50 mm1001 * 100 items£163.00: HAWP05000: MILFHAWP05000
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP09000HAWP09000HAWP, white, plain0,45 µm90 mm1001 * 100 items£415.00: HAWP09000: MILFHAWP09000
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFVSWP01300VSWP01300VSWP, white, plain0,025 µm13 mm1001 * 100 items£213.00: VSWP01300: MILFVSWP01300
Specifications
TypeVSWP, white, plainEnvironmentally PreferablePore size0,025 µmThickness105 µmWater flow rate0,15 ml/min/cm²Airflow rate0,15 L/min/cm²Max. temperature75 °C - MILFAAWP09000AAWP09000AAWP, white, plain0,80 µm90 mm1001 * 100 items£370.00: AAWP09000: MILFAAWP09000
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFHAWP04700F1HAWP04700F1HAWP, white, plain0,45 µm47 mm1001 * 100 items£461.00: HAWP04700F1: MILFHAWP04700F1
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness150 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFRAWP05000RAWP05000RAWP, white, plain1,20 µm50 mm1001 * 100 items£196.00: RAWP05000: MILFRAWP05000
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness150 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - 512-0241AAWP04700AAWP, white, plain0,80 µm47 mm1001 * 100 items£161.00: AAWP04700: MILFAAWP04700
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWP0470MAAWP0470MAAWP, white, plain0,80 µm47 mm501 * 50 items£615.00: AAWP0470M: MILFAAWP0470M
Specifications
TypeAAWP, white, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWP037P0AAWP037P0AAWP, white, plain, with thick pad0,80 µm37 mm1001 * 100 items£170.00: AAWP037P0: MILFAAWP037P0
Specifications
TypeAAWP, white, plain, with thick padEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWP037PMAAWP037PMAAWP, white, plain, with thick pad0,80 µm37 mm501 * 50 items£557.00: AAWP037PM: MILFAAWP037PM
Specifications
TypeAAWP, white, plain, with thick padEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWP03700AAWP03700AAWP, white, plain, with thin pad0,80 µm37 mm1001 * 100 items£149.00: AAWP03700: MILFAAWP03700
Specifications
TypeAAWP, white, plain, with thin padEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFDAWP02500DAWP02500DAWP, white, plain0,65 µm25 mm1001 * 100 items£115.00: DAWP02500: MILFDAWP02500
Specifications
TypeDAWP, white, plainEnvironmentally PreferablePore size0,65 µmThickness180 µmWater flow rate140 ml/min/cm²Airflow rate9 L/min/cm²Max. temperature75 °C - MILFDAWP09025DAWP09025DAWP, white, plain0,65 µm90 mm1001 * 100 items£127.00: DAWP09025: MILFDAWP09025
Specifications
TypeDAWP, white, plainEnvironmentally PreferablePore size0,65 µmThickness180 µmWater flow rate140 ml/min/cm²Airflow rate9 L/min/cm²Max. temperature75 °C - MILFDAWP14250DAWP14250DAWP, white, plain0,65 µm142 mm501 * 50 items£412.00: DAWP14250: MILFDAWP14250
Specifications
TypeDAWP, white, plainEnvironmentally PreferablePore size0,65 µmThickness180 µmWater flow rate140 ml/min/cm²Airflow rate9 L/min/cm²Max. temperature75 °C - 512-0250GSWP02500GSWP, white, plain0,22 µm25 mm1001 * 100 items£115.00: GSWP02500: MILFGSWP02500
Specifications
TypeGSWP, white, plainEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - 512-0251GSWP04700GSWP, white, plain0,22 µm47 mm1001 * 100 items£169.00: GSWP04700: MILFGSWP04700
Specifications
TypeGSWP, white, plainEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFHAEP04700HAEP04700White, plain0,45 µm47 mm1001 * 100 items£261.00: HAEP04700: MILFHAEP04700
Specifications
TypeWhite, plainEnvironmentally PreferablePore size0,45 µmThickness150 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWP14250HAWP14250HAWP, white, plain0,45 µm142 mm501 * 50 items£375.00: HAWP14250: MILFHAWP14250
Specifications
TypeHAWP, white, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFPHWP02500PHWP02500PHWP, white, plain0,30 µm25 mm1001 * 100 items£163.00: PHWP02500: MILFPHWP02500
Specifications
TypePHWP, white, plainEnvironmentally PreferablePore size0,30 µmThickness170 µmWater flow rate32 ml/min/cm²Airflow rate3 L/min/cm²Max. temperature75 °C - MILFPHWP09025PHWP09025PHWP, white, plain0,30 µm90 mm251 * 25 items£171.00: PHWP09025: MILFPHWP09025
Specifications
TypePHWP, white, plainEnvironmentally PreferablePore size0,30 µmThickness170 µmWater flow rate32 ml/min/cm²Airflow rate3 L/min/cm²Max. temperature75 °C - MILFPHWP14250PHWP14250PHWP, white, plain0,30 µm142 mm501 * 50 items£500.00: PHWP14250: MILFPHWP14250
Specifications
TypePHWP, white, plainEnvironmentally PreferablePore size0,30 µmThickness170 µmWater flow rate32 ml/min/cm²Airflow rate3 L/min/cm²Max. temperature75 °C - MILFRAWP02500RAWP02500RAWP, white, plain1,20 µm25 mm1001 * 100 items£116.00: RAWP02500: MILFRAWP02500
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - 512-0267RAWP04700RAWP, white, plain1,20 µm47 mm1001 * 100 items£168.00: RAWP04700: MILFRAWP04700
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFRAWP09025RAWP09025RAWP, white, plain1,20 µm90 mm251 * 25 items£141.00: RAWP09025: MILFRAWP09025
Specifications
TypeRAWP, white, plainEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFSCWP01300SCWP01300SCWP, white, plain8,00 µm13 mm1001 * 100 items£94.40: SCWP01300: MILFSCWP01300
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - 512-0270SCWP02500SCWP, white, plain8,00 µm25 mm1001 * 100 items£116.00: SCWP02500: MILFSCWP02500
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - 512-0271SCWP04700SCWP, white, plain8,00 µm47 mm1001 * 100 items£168.00: SCWP04700: MILFSCWP04700
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - MILFSCWP09025SCWP09025SCWP, white, plain8,00 µm90 mm251 * 25 items£139.00: SCWP09025: MILFSCWP09025
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - MILFSCWP14250SCWP14250SCWP, white, plain8,00 µm142 mm501 * 50 items£398.00: SCWP14250: MILFSCWP14250
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - MILFSCWP0190RSCWP0190RSCWP, white, plain8,00 µm19×42 mm1001 * 100 items£202.00: SCWP0190R: MILFSCWP0190R
Specifications
TypeSCWP, white, plainEnvironmentally PreferablePore size8,00 µmThickness160 µmWater flow rate620 ml/min/cm²Airflow rate65 L/min/cm²Max. temperature75 °C - 512-0273SMWP01300SMWP, white, plain5,00 µm13 mm1001 * 100 items£96.70: SMWP01300: MILFSMWP01300
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - 512-0274SMWP02500SMWP, white, plain5,00 µm25 mm1001 * 100 items£119.00: SMWP02500: MILFSMWP02500
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - 512-0275SMWP04700SMWP, white, plain5,00 µm47 mm1001 * 100 items£168.00: SMWP04700: MILFSMWP04700
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - MILFSMWP0190RSMWP0190RSMWP, white, plain5,00 µm19×42 mm1001 * 100 items£202.00: SMWP0190R: MILFSMWP0190R
Specifications
TypeSMWP, white, plainEnvironmentally PreferablePore size5,00 µmThickness160 µmWater flow rate580 ml/min/cm²Airflow rate32 L/min/cm²Max. temperature75 °C - 512-0276SSWP02500SSWP, white, plain3,00 µm25 mm1001 * 100 items£119.00: SSWP02500: MILFSSWP02500
Specifications
TypeSSWP, white, plainEnvironmentally PreferablePore size3,00 µmThickness180 µmWater flow rate320 ml/min/cm²Airflow rate30 L/min/cm²Max. temperature75 °C - MILFSSWP09025SSWP09025SSWP, white, plain3,00 µm90 mm251 * 25 items£134.00: SSWP09025: MILFSSWP09025
Specifications
TypeSSWP, white, plainEnvironmentally PreferablePore size3,00 µmThickness180 µmWater flow rate320 ml/min/cm²Airflow rate30 L/min/cm²Max. temperature75 °C - MILFSSWP14250SSWP14250SSWP, white, plain3,00 µm142 mm501 * 50 items£397.00: SSWP14250: MILFSSWP14250
Specifications
TypeSSWP, white, plainEnvironmentally PreferablePore size3,00 µmThickness180 µmWater flow rate320 ml/min/cm²Airflow rate30 L/min/cm²Max. temperature75 °C - MILFVCWP01300VCWP01300VCWP, white, plain0,10 µm13 mm1001 * 100 items£214.00: VCWP01300: MILFVCWP01300
Specifications
TypeVCWP, white, plainEnvironmentally PreferablePore size0,10 µmThickness105 µmWater flow rate1,5 ml/min/cm²Airflow rate0,4 L/min/cm²Max. temperature75 °C - MILFVCWP02500VCWP02500VCWP, white, plain0,10 µm25 mm1001 * 100 items£288.00: VCWP02500: MILFVCWP02500
Specifications
TypeVCWP, white, plainEnvironmentally PreferablePore size0,10 µmThickness105 µmWater flow rate1,5 ml/min/cm²Airflow rate0,4 L/min/cm²Max. temperature75 °C - MILFVCWP09025VCWP09025VCWP, white, plain0,10 µm90 mm251 * 25 items£260.00: VCWP09025: MILFVCWP09025
Specifications
TypeVCWP, white, plainEnvironmentally PreferablePore size0,10 µmThickness105 µmWater flow rate1,5 ml/min/cm²Airflow rate0,4 L/min/cm²Max. temperature75 °C - MILFVCWP14250VCWP14250VCWP, white, plain0,10 µm142 mm501 * 50 items£561.00: VCWP14250: MILFVCWP14250
Specifications
TypeVCWP, white, plainEnvironmentally PreferablePore size0,10 µmThickness105 µmWater flow rate1,5 ml/min/cm²Airflow rate0,4 L/min/cm²Max. temperature75 °C - MILFVMWP01300VMWP01300VMWP, white, plain0,05 µm13 mm1001 * 100 items£236.00: VMWP01300: MILFVMWP01300
Specifications
TypeVMWP, white, plainEnvironmentally PreferablePore size0,05 µmThickness105 µmWater flow rate0,74 ml/min/cm²Airflow rate0,25 L/min/cm²Max. temperature75 °C - MILFVMWP02500VMWP02500VMWP, white, plain0,05 µm25 mm1001 * 100 items£273.00: VMWP02500: MILFVMWP02500
Specifications
TypeVMWP, white, plainEnvironmentally PreferablePore size0,05 µmThickness105 µmWater flow rate0,74 ml/min/cm²Airflow rate0,25 L/min/cm²Max. temperature75 °C - 512-0281VMWP04700VMWP, white, plain0,05 µm47 mm1001 * 100 items£359.00: VMWP04700: MILFVMWP04700
Specifications
TypeVMWP, white, plainEnvironmentally PreferablePore size0,05 µmThickness105 µmWater flow rate0,74 ml/min/cm²Airflow rate0,25 L/min/cm²Max. temperature75 °C - MILFVMWP09025VMWP09025VMWP, white, plain0,05 µm90 mm251 * 25 items£322.00: VMWP09025: MILFVMWP09025
Specifications
TypeVMWP, white, plainEnvironmentally PreferablePore size0,05 µmThickness105 µmWater flow rate0,74 ml/min/cm²Airflow rate0,25 L/min/cm²Max. temperature75 °C - 512-0282VSWP02500VSWP, white, plain0,025 µm25 mm1001 * 100 items£288.00: VSWP02500: MILFVSWP02500
Specifications
TypeVSWP, white, plainEnvironmentally PreferablePore size0,025 µmThickness105 µmWater flow rate0,15 ml/min/cm²Airflow rate0,15 L/min/cm²Max. temperature75 °C
Standard MF-Millipore™ membranes, white, gridded
- TypePore sizeØPk
- MILFRAWG04700RAWG04700RAWG, white, gridded1,20 µm47 mm1001 * 100 items£182.00: RAWG04700: MILFRAWG04700
Specifications
TypeRAWG, white, griddedEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFRAWG02500RAWG02500RAWG, white, gridded1,20 µm25 mm1001 * 100 items£142.00: RAWG02500: MILFRAWG02500
Specifications
TypeRAWG, white, griddedEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFHAWG01300HAWG01300HAWG, white, gridded0,45 µm13 mm1001 * 100 items£114.00: HAWG01300: MILFHAWG01300
Specifications
TypeHAWG, white, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFAAWG04700AAWG04700AAWG, white, gridded0,80 µm47 mm1001 * 100 items£169.00: AAWG04700: MILFAAWG04700
Specifications
TypeAAWG, white, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWG03700AAWG03700AAWG, white, gridded, with thin pad0,80 µm37 mm1001 * 100 items£189.00: AAWG03700: MILFAAWG03700
Specifications
TypeAAWG, white, gridded, with thin padEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWG0250CAAWG0250CAAWG, white, gridded0,80 µm25 mm1001 * 100 items£134.00: AAWG0250C: MILFAAWG0250C
Specifications
TypeAAWG, white, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWG02500AAWG02500AAWG0,80 µm25 mm1001 * 100 items£124.00: AAWG02500: MILFAAWG02500
Specifications
TypeAAWGEnvironmentally PreferablePore size0,80 µmThickness150 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAAWG01300AAWG01300AAWG, white, gridded0,80 µm13 mm1001 * 100 items£115.00: AAWG01300: MILFAAWG01300
Specifications
TypeAAWG, white, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFHAWG04700HAWG04700HAWG, white, gridded0,45 µm47 mm1001 * 100 items£170.00: HAWG04700: MILFHAWG04700
Specifications
TypeHAWG, white, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWG05000HAWG05000HAWG, white, gridded0,45 µm50 mm2001 * 200 items£215.00: HAWG05000: MILFHAWG05000
Specifications
TypeHAWG, white, griddedEnvironmentally PreferablePore size0,45 µmThickness-Water flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFRAWG0250CRAWG0250CRAWG, white, gridded1,20 µm25 mm1001 * 100 items£147.00: RAWG0250C: MILFRAWG0250C
Specifications
TypeRAWG, white, griddedEnvironmentally PreferablePore size1,20 µmThickness170 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFHAWG03700HAWG03700HAWG, white, gridded, with thin pad0,45 µm37 mm1001 * 100 items£200.00: HAWG03700: MILFHAWG03700
Specifications
TypeHAWG, white, gridded, with thin padEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWG02500HAWG02500HAWG, white, gridded0,45 µm25 mm1001 * 100 items£124.00: HAWG02500: MILFHAWG02500
Specifications
TypeHAWG, white, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHAWG04705HAWG04705HAWG, white, gridded0,45 µm47 mm5001 * 500 items£518.00: HAWG04705: MILFHAWG04705
Specifications
TypeHAWG, white, griddedEnvironmentally PreferablePore size0,45 µmThickness150 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C
Standard MF-Millipore™ membranes, black, plain
- TypePore sizeØPk
- MILFHABP04700HABP04700HABP, black, plain0,45 µm47 mm1001 * 100 items£212.00: HABP04700: MILFHABP04700
Specifications
TypeHABP, black, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHABP02500HABP02500HABP, black, plain0,45 µm25 mm1001 * 100 items£163.00: HABP02500: MILFHABP02500
Specifications
TypeHABP, black, plainEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFAABPC5000AABPC5000AABP, black, plain0,8 µm50 mm11 * 1 items£183.00: AABPC5000: MILFAABPC5000
Specifications
TypeAABP, black, plainEnvironmentally PreferablePore size0,8 µmThickness—Water flow rate—Airflow rate—Max. temperature— - MILFAABP04700AABP04700AABP, black, plain0,80 µm47 mm1001 * 100 items£212.00: AABP04700: MILFAABP04700
Specifications
TypeAABP, black, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAABP02500AABP02500AABP, black, plain0,80 µm25 mm1001 * 100 items£168.00: AABP02500: MILFAABP02500
Specifications
TypeAABP, black, plainEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C
Standard MF-Millipore™ membranes, black, gridded
- TypePore sizeØPk
- MILFAABG04700AABG04700AABG, black, gridded0,80 µm47 mm1001 * 100 items£229.00: AABG04700: MILFAABG04700
Specifications
TypeAABG, black, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAABG03700AABG03700AABG, black, gridded, with thin pad0,80 µm37 mm1001 * 100 items£211.00: AABG03700: MILFAABG03700
Specifications
TypeAABG, black, gridded, with thin padEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAABG02500AABG02500AABG, black, gridded0,80 µm25 mm1001 * 100 items£165.00: AABG02500: MILFAABG02500
Specifications
TypeAABG, black, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFAABG01300AABG01300AABG, black, gridded0,80 µm13 mm1001 * 100 items£169.00: AABG01300: MILFAABG01300
Specifications
TypeAABG, black, griddedEnvironmentally PreferablePore size0,80 µmThickness180 µmWater flow rate190 ml/min/cm²Airflow rate16 L/min/cm²Max. temperature75 °C - MILFHABG02500HABG02500HABG, black, gridded0,45 µm25 mm1001 * 100 items£192.00: HABG02500: MILFHABG02500
Specifications
TypeHABG, black, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHABG01300HABG01300HABG, black, gridded0,45 µm13 mm1001 * 100 items£163.00: HABG01300: MILFHABG01300
Specifications
TypeHABG, black, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C - MILFHABG04700HABG04700HABG, black, gridded0,45 µm47 mm1001 * 100 items£232.00: HABG04700: MILFHABG04700
Specifications
TypeHABG, black, griddedEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rate60 ml/min/cm²Airflow rate4 L/min/cm²Max. temperature75 °C
MF-Millipore™ membranes without Triton® surfactant, white, plain
- TypePore sizeØPk
- MILFHATF01300HATF01300HATF, white, plain, Triton®-free0,45 µm13 mm1001 * 100 items£113.00: HATF01300: MILFHATF01300
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFHATF09025HATF09025HATF, white, plain, Triton®-free0,45 µm90 mm251 * 25 items£155.00: HATF09025: MILFHATF09025
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFHATF10050HATF10050HATF, white, plain, Triton®-free0,45 µm100 mm1001 * 100 items£331.00: HATF10050: MILFHATF10050
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFHATF14250HATF14250HATF, white, plain, Triton®-free0,45 µm142 mm501 * 50 items£431.00: HATF14250: MILFHATF14250
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFHATF02500HATF02500HATF, white, plain, Triton®-free0,45 µm25 mm1001 * 100 items£131.00: HATF02500: MILFHATF02500
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFGSTF04700GSTF04700GSTF, white, plain, Triton®-free0,22 µm47 mm1001 * 100 items£168.00: GSTF04700: MILFGSTF04700
Specifications
TypeGSTF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFGSTF09000GSTF09000GSTF, white, plain, Triton®-free0,22 µm90 mm1001 * 100 items£493.00: GSTF09000: MILFGSTF09000
Specifications
TypeGSTF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFHATF08250HATF08250HATF, white, plain, Triton®-free0,45 µm82 mm501 * 50 items£262.00: HATF08250: MILFHATF08250
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - MILFGSTF02500GSTF02500GSTF, white, plain, Triton®-free0,22 µm25 mm1001 * 100 items£120.00: GSTF02500: MILFGSTF02500
Specifications
TypeGSTF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFHATF04700HATF04700HATF, white, plain, Triton®-free0,45 µm47 mm1001 * 100 items£202.00: HATF04700: MILFHATF04700
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature - : RATF14250: MILFRATF14250
Specifications
TypeRATF, white, plain, Triton®-freeEnvironmentally PreferablePore size1,20 µmThickness180 µmWater flow rate270 ml/min/cm²Airflow rate20 L/min/cm²Max. temperature75 °C - MILFGSTF01300GSTF01300GSTF, white, plain, Triton®-free0,22 µm13 mm1001 * 100 items£107.00: GSTF01300: MILFGSTF01300
Specifications
TypeGSTF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFGSTF14250GSTF14250GSTF, white, plain, Triton®-free0,22 µm142 mm501 * 50 items£402.00: GSTF14250: MILFGSTF14250
Specifications
TypeGSTF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,22 µmThickness180 µmWater flow rate18 ml/min/cm²Airflow rate2 L/min/cm²Max. temperature75 °C - MILFHATF08550HATF08550HATF, white, plain, Triton®-free0,45 µm85 mm501 * 50 items£262.00: HATF08550: MILFHATF08550
Specifications
TypeHATF, white, plain, Triton®-freeEnvironmentally PreferablePore size0,45 µmThickness180 µmWater flow rateAirflow rateMax. temperature