Power supply, PS300B
Supplier: Hoefer
732-1695EA 796 GBP
HOEFPS300B
Power supply, PS300B
The versatile PS300B power supply is designed for most electrophoresis techniques.
  • Constant voltage or constant current mode with automatic crossover
  • Four sets of 4 mm output jacks
  • Continuous runs or 999 min timer
  • Automatic recovery after power failure
  • Open circuit detection
  • Short circuit protection