(±)-4-HDoHE
Supplier: Cayman Chemical
CAYM33200-100EA
457
GBP
CAYM33200-100
CAYM33200-25
CAYM33200-250
CAYM33200-50
(±)-4-HDoHE
(±)-4-HDoHE
Formula:
C₂₂H₃₂O₃ |
CAS Number:
90906-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...