Northern Ireland
Order Entry
Methyl-β-D-glucopyranoside hemihydrate 99%
Irritant
Methyl-β-D-glucopyranoside hemihydrate 99%
Catalog # ACRO270920250
CAS Number:  
undefined
Irritant
Methyl-β-D-glucopyranoside hemihydrate 99%
Catalog # ACRO270920250
CAS Number:  
Price
£497.00
Log in for availability
Unit of Measure 1 * 25 g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤5.4 % (K.F.)
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥98.5 % (ELSD)
  • Specific optical rotation:
    -31° to -35° (20°C, 589 nm) (c=4, water)

Specification Test Results

About this item

 :
 :203.19 g/mol
Melting Pt:111…113 °C
EINECS:211-909-9
 :GHS07
Storage Temperature:Refrigerator
CAS Number:7000-27-3
 :709-50-2
Formula:C₇H₁₄O₆·0.5H₂O
MDL Number:MFCD00149442