VWR®, Bradford protein assay
Supplier: VWR
E535-KITEA 167 GBP
VWRCE535-KIT
VWR®, Bradford protein assay
The Bradford Protein Assay is the preferred colorimetric assay for quantifying total protein concentration. Based upon complex formation between basic and aromatic amino acid residues with Coomassie® Brilliant Blue G-250 dye, the Bradford method is easier, faster and more sensitive than the Lowry method.
  • Sensitive, fast and consistent
  • Standard protocol for measuring concentrations from 10 to 100 μg
  • Microassay protocol for measuring concentrations from 1 to 10 μg
Delivery information: Kit includes Bradford reagent, 1 L; and bovine albumin standard, 0,5 mg/ml, 1,5 ml.