Headtops for respiratory system Versaflo™, M-100 series
Supplier: 3M
MMMAM107EA 358 GBP
MMMAM107
Headtops for respiratory system Versaflo™, M-100 series
These face shields combine respiratory protection with eye and face protection, and with some models, head and hearing protection. The face shields are designed to be used with an approved 3M air delivery unit to form a respiratory protective device. All face shields have a fully adjustable suspension for optimal fit and weight distribution. They incorporate a deflector to allow the user to direct the airflow inside the face shield for increased control and comfort.
  • M-106:
  • Comfortable PU coated polyamide faceseal; coated PC visor
  • Hazard type: Gas, vapour and particulates, liquid splash, low energy flying particles, dust
  • Protection level (NPF): 50 (Jupiter/TR-302E/TR-602E/V-Series)
  • Used for spraying, chemical processing or in construction
  • M-107:
  • Polyester faceseal; coated PC visor
  • With flame resistant faceseal for applications involving hot particles
  • Hazard type: Gas, vapour and particulates, liquid splash, low energy flying particles, dust
  • Protection level (NPF): 50 (Jupiter/TR-302E/TR-602E/V-Series)
  • Used for metal working, chemical processing and pharmaceutical manufacturing
Certifications: M-106: EN 166:2001, EN 12941 TH2, EN 14594 2B
M-107: EN 12941 TH2, EN 14594 2B