Cleanroom hoods, STH002
Supplier: Ansell-Nitritex
NITRSTH002-C40-MEA 0 GBP
NITRSTH002-C40-M NITRSTH002-C40-L
Cleanroom hoods, STH002
These cleanroom hoods are made of MT031 fabric consisting of 99% polyester and 1% carbon.
These cleanroom hoods are made of MT031 fabric consisting of 99% polyester and 1% carbon.
  • Front with stud fastening
  • Horizontal stud back
  • Vertical stud back
  • Suitable for irradiation
Suitable for use in Class 10, ISO 4, M2,5 and above cleanrooms.
  • Front with stud fastening
  • Horizontal stud back
  • Vertical stud back
  • Suitable for irradiation
109 g/m²