Aerator for BOD bottles, WHEATON®
Supplier: DWK Life Sciences
WHEA227700EA 241 GBP
WHEA227700
Aerator for BOD bottles, WHEATON®
Low extractable borosilicate glass aerator for use with 300 ml bottles with glass frit.
  • Glass frit porosity (40 to 60 μm)
  • EPA method 245.1 (Manual cold vapor technique)
Used in determining inorganic forms of mercury and organic mercurial in drinking, surface and saline waters, as well as domestic and industrial waste.
Certifications: Material conforms to USP Type I and ASTM E 438 Type I, Class A requirements.