Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Northern Ireland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
£0.00
Order Entry
tert-Butylcyclohexane ≥99%
:
Thermo Fisher Scientific
tert-Butylcyclohexane ≥99%
:
Thermo Fisher Scientific
:
3178-22-1
:
:
C₁₀H₂₀
:
MFCD00001471
:
140.27 g/mol
:
171 °C
:
–41 °C
:
0,830
:
40 °C (104 °F)
:
Ambient
:
221-652-4
:
3295
:
3,III
:
GHS02
msds_ALFAB24126
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Pack type
Pk
B24126.14
B24126.14
Glass bottle for solids
25 g
1 * 25 g
1 * 25 g - £54.50
£54.50
-
+
1 * 25 g
£54.50
-
+
B24126.22
B24126.22
Glass bottle for solids
100 g
1 * 100 g
1 * 100 g - £187.00
£187.00
-
+
1 * 100 g
£187.00
-
+