Saccharin, Supelco®

Supplier: Merck

Synonyms: o-Sulphobenzimideo-Benzoic sulphimide

41131-5GEA 63 GBP
41131-5G
Saccharin, Supelco®
Saccharin
Formula: C₇H₅NO₃S
MW: 183,19 g/mol
MDL Number: MFCD00005866
CAS Number: 81-07-2
EINECS: 201-321-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR