Northern Ireland
Order Entry
Supelco® Organic Reference Standard, Ammonium sulfate
Supelco® Organic Reference Standard, Ammonium sulfate
  59845-25G
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Ammonium sulfate
  59845-25G
 :  Merck
 :  
 
£55.00
   
  1 * 25 g

 

  • Pk:
    25 g
  • CAS No.:
    7783-20-2
  • Description:
    Ammonium sulfate
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003391
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Mascagnite
  • Melting point:
    >280 °C (dec.) (lit.)
  • Formula:
    (NH₄)₂SO₄
  • Molecular weight:
    132,14
  • Density:
    1,77 g/ml at 25 °C (lit.)