Northern Ireland
Order Entry
1-Iodo-2-nitrobenzene 97%
Irritant
1-Iodo-2-nitrobenzene 97%
Catalog # ACRO159450250
CAS Number:  
undefined
Irritant
1-Iodo-2-nitrobenzene 97%
Catalog # ACRO159450250
CAS Number:  
Price
£151.00
Log in for availability
Unit of Measure 1 * 25 g

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Yellow-green to yellow-brown Granular powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 52 °C
  • GC:
    ≥96.0 %
  • Oxidizing efficiency:
    ≥99.6 %

Specification Test Results

About this item

 :
 :249.01 g/mol
Boiling Pt:289 °C (1013 hPa)
Melting Pt:47…52 °C
Density:1,9186 g/cm³ (75 °C)
Flash Pt:110 °C
Storage Temperature:Refrigerator
EINECS:210-200-1
 :GHS07
CAS Number:609-73-4
Formula:C₆H₄INO₂
MDL Number:MFCD00007088