Safety funnels with lid
Supplier: SCAT Europe
221-0198EA 189 GBP
SCTV118964
Safety funnels with lid
HDPE, black, electroconductive with grounding dipper.
  • Removable sieves for cleaning
  • Low installed height, even fits below low work surfaces
  • Various thread diameters
Suitable for use with all types of chemicals. The screw cap rotates freely, which makes it easier to screw the funnel on and off. 135 mm Ø without lance.