n-Octacosane, Supelco®
Supplier: Merck
Synonyms:
N-octacosane
M-Clarity™ quality level = MQ100
74684-250MGEA
63.4
GBP
74684-250MG
n-Octacosane, Supelco®
n-Octacosane
Formula:
H₃C(CH₂)₂₆CH₃ MW: 394,77 g/mol Boiling Pt: 432 °C (1013 hPa) Melting Pt: 60…61 °C Density: 0,8067 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 EINECS: 211-125-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...