Bromoform ≥98.0% stabilised, AnalaR NORMAPUR® analytical reagent, for mineralogy
|
Danger
|
Synonyms:
Tribromomethane
Stabilisation:
Stabilised with ethanol 1 %
22045.297EA
717
GBP
22045.297
Bromoform ≥98.0% stabilised, AnalaR NORMAPUR® analytical reagent, for mineralogy
Bromoform
Formula:
Br₃CH MW: 252,73 g/mol Boiling Pt: 149,5 °C (1013 hPa) Melting Pt: 9,2 °C Density: 2,8912 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000128 CAS Number: 75-25-2 EINECS: 200-854-6 UN: 2515 ADR: 6.1,III Merck Index: 13,01404 |
Specification Test Results
Assay (on anhydrous substance) | Min. 98.0 % |
Acidity | Max. 0.002 meq/g |
Density (20/4) | 2.820 - 2.880 |
Refractive index (20°C) | 1.590 - 1.598 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...