Lithium chloride monohydrate, Suprapur® for trace analysis, Supelco®

Warning

Synonyms: Lithium chloride hydrateHydrochloric acid lithium salt monohydrate

M-Clarity™ quality level = MQ100

1.05677.0250EA 1020 GBP
1.05677.0250
Lithium chloride monohydrate, Suprapur® for trace analysis, Supelco®
Lithium chloride monohydrate
Formula: ClLi·1H₂O
MW: 60,41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 16712-20-2
EINECS: 231-212-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 614 °C (anhydrous substance)
Molar Mass 60.41 g/mol
Bulk Density 600 kg/m3
Boiling Point 1360 °C (anhydrous substance)
Density 5.5 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR