Chamber slides, BioCoat™
Supplier: Corning Life Sciences
734-1070EA 0 GBP
BDAA354656
Chamber slides, BioCoat™
Chamber slides with a selection of different extracellular matrix proteins and attachment factors.
  • Glass with PS vessel, lid and safety removal tool
  • Different coatings: Collagen I, Fibronectin or Poly-D-Lysine
  • For determining the extracellular matrix for cell types and applications