You Searched For: Shaking Water Baths


11  results were found

SearchResultCount:"11"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Collection
Description: Linear shaking water baths with stainless steel tank and simple, intuitive user interface. It is quick and easy to fit alternative shaking trays.

Supplier: JULABO GmbH
Description: Stainless steel shaking water baths are suitable for unsupervised continuous operation. The bright, LED Multi-Display showing up to six temperature va...

Supplier: GRANT INSTRUMENTS LTD.
Description: The Aqua Pro series of shaking water baths offers two bath models with exceptional temperature performance and usability. The baths are ideal for basi...

Catalog Number: (462-0519)
Supplier: GRANT INSTRUMENTS LTD.
Description: The OLS shaking water bath offers exceptional temperature performance and usability. The bath is ideal for basic as well as more demanding application...
UOM: 1 * 1 items


Supplier: LAUDA
Description: Shaking water baths for exactly reproducible temperatures. The working temperature range is 25 to 100 °C.

Supplier: NICKEL ELECTRO
Description: Digital and analogue linear shaking water baths with a stainless steel tank housed in a durable outer case. The corrosion resistant reciprocating trol...

Catalog Number: (GERB3550)
Supplier: GERBER FUNKE DR N.
Description: Shaking water bath, stainless steel. PID controller with PT-100 temperature sensor.
UOM: 1 * 1 items


Catalog Number: (EDVO5027)
Supplier: EDVOTEK EUROPE LTD
Description: Digital stainless steel shaking water bath, designed for when incubation of samples together with precise aeration and agitation is required.
UOM: 1 * 1 items


Catalog Number: (HAKATSCIR35)
Supplier: THERMO ELECTRON LED GMBH
Description: These circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in se...
UOM: 1 * 1 items


Supplier: THERMO ELECTRON LED GMBH
Description: Precisionâ„¢ water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetic...

Catalog Number: (STUA12152-02)
Supplier: Cole Parmer
Description: Customisethe shaking water bath to your requirements with platforms and racks
UOM: 1 * 1 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 11 of 11
no targeter for Bottom