Chemically resistant vacuum system, SR 840
Supplier: KNF
181-3402EA 2770 GBP
KNFASR820
Chemically resistant vacuum system, SR 840
Model SR 840
This vacuum system is equipped with a chemically resistant vacuum pump N 840.3 FT.18 (for a full description, see 189-0017). The SR 840 includes two separators and can be used in a wide range of laboratory tasks wherever a clean and constant vacuum is needed.
  • Resistant to aggressive gases and vapours
  • Solvent droplets in the gas are separated pressure/exhaust side
  • Particles are trapped in the separators (suction side)
  • Maintenance-free, quiet running, easy to upgrade and retrofit
Delivery information: Includes the chemically resistant diaphragm pump with base plate, two separators for the suction and pressure sides, with hoses.