Orbital platform shakers, Labline MaxQ™ 2000 & 3000
Supplier: Thermo Scientific
BARNSHKE2000GB-1CEEA 4940 GBP
BARNSHKE2000GB-1CE BARNSHKE2000GA-1CE BARNSHKE3000-1CE BARNSHKE2000-1CE
Orbital platform shakers, Labline MaxQ™ 2000 & 3000
Versatile units offering several platform sizes with the ability to shake multiple vessel configurations.
  • Choice of analogue and digital models with five year warranty on parts and LIFETIME on drive mechanism
  • Continuous or timed operation for analogue models 1 to 60 min, digital models 0,1 to 999 min or 0,1 up to 999 h
  • Triple eccentric drive enables continuous 24 hour operation, handles heavier loads and provides smooth uniform agitation of full or unbalanced loads even at high speeds
  • Can be used at 0 to 40 °C (20 to 80% relative humidity) in incubators, warm rooms, environmental chambers and refrigeratorS
Analogue Operating System:
DC motor and variable speed control from 40 to 400 min⁻¹, controlled by a rotary dial. An integrated tachometer, monitors and displays speed in rpm to guarantee accurate setting.
Digital Operating System:
Units have a solid-state DC brushless motor, with variable speed control from 15 to 500 min⁻¹. The time display shows time counting up during continuous operation and down during timed runs. It is possible to view speed and operating time simultaneously on 2 separate LED displays, which helps minimise errors in establishing running conditions. The shaker will shut down and audible/visual alarms signal if unit operates 10% over/unset speed, preventing shaker from moving on the bench top. The soft start feature eliminates splashing of vessel contents, wetting of flask closure, and helps prevent sudden starts and stops. Set point retention retains parameters during power failure and restarts the unit automatically after power is restored. RS232 interface allows remote monitoring of speed and time.
Packages are available with genomic application which includes shaker, platform and clamps.
Ordering information: All platforms and related accessories must be ordered separately. Clamps for universal platform: Maximum number of clamps per platform size (mm) as shown below.