Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Ice Crushers

https://ni.vwr.com/stibo/smallweb/std.lang.all/94/20/18989420.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (474-0044)
Supplier:  WARING BLENDOR
Description:   The ice crusher is portable, of compact design and especially suitable for countertop use. It has a shaded pole gear motor, 24 rpm. The special height...
UOM:  1 * 1 items
Catalog Number: (474-0029)
Supplier:  SCOTSMAN
Description:   The Crushman 360 is manufactured from stainless steel.
UOM:  1 * 1 items
Catalog Number: (HOSHC-105)
Supplier:  HOSHIZAKI
Description:   Ice crushers are ideal for crushing large quantities of ice cubes for perfectly presented drinks.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3  of 3