Refrigerated centrifuge for large-volume centrifugation (4×1 litre) or centrifugation of a large number of samples (160×blood sample tubes, 96×15 ml Falcon® conical tubes). Up to 32 standard microplates can be centrifuged in a single cycle.
- Easy-to-clean touch keyboard
- Low-maintenance, brushless, induction motor
- Optimum stability thanks to SMARTSpinTM Imbalance Detection System
- Wide range of rotors available: LAC-250 fixed angle rotor; LH-4000 swing-out rotor; Diagnostik™ rotor suitable for the most commonly used clinical analysis tube racks; highly versatile LH-4000 swing-out rotor (for tissue culture, virological tests, blood treatment etc.); and LH-4000W swing-out rotor specially designed for high-speed, low-temperature protocols, which maintains the required conditions for temperature-sensitive samples
Certifications: EN 61 010-1, EN 61 010-2-020, EN 50 081-1 and EN 50 082-1 compliant