BOD measurement, OxiTop® Control 6
Supplier: WTW
OxiTop®
705-1000EA 5540 GBP
WTWA208201
BOD measurement, OxiTop® Control 6
The automated system for measuring BOD, OxiTop® Control, consists of an OxiTop® Control measuring head and an infrared controller. This system is exceptionally easy to use. The controller is capable of configuring up to 100 measuring heads and receiving the measurement data via infrared transmission. Each sample is easily identified and recognised by the controller thanks to the measuring head, each of which is associated with a unique serial number. The controller is multilingual to make it as easy as possible to operate. The digital display is clear and offers step by step guidance for all user operations.
  • Standard and routine measurements of 6 or 12 samples in parallel with automatic data transfer
  • Controller has a range of between 0...40/80/200/400/800/2000/4000 mg/l BOD
  • Automatic assignment of a sample identification number
  • Measurement period: 0,5 h to 99 days
  • Heads can store up to 360 data sets
OxiTop® Control 6
Complete package for 6 simultaneous measurements with OxiTop® OC 100 Controller, and IS 6 Inductive Stirring System for mains operation 230 V/50/60 Hz and 6 OxiTop® C measuring heads and appropriate standard accessories.
Delivery information: Delivered with brown sample bottles, stirrer bars, stirrer bar retriever, rubber sleeves, NaOH pellets, 1 each of overflow measuring flask 164 ml and 432 ml, nitrification inhibitor.