Environmentally friendly thermometers, enviro safe
620-0880EA 16.6 GBP
VWRI620-0880
Environmentally friendly thermometers, enviro safe
These earth friendly thermometers feature green, non toxic, biodegradable, enviro safe liquid against lead-free, white back glass. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Meet government, education, and industry mandates to purchase 'Green' products
  • Individually serialised
Certifications: Traceable to NIST. Envirokleen™ certified by ChemTel, an independent laboratory.
Delivery information: Includes a multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability.