Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Northern Ireland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
£0.00
Order Entry
Home
PPE Dispensers
VWR®, Shoe Cover Dispenser
VWR®, Shoe Cover Dispenser
VWR®, Shoe Cover Dispenser
113-0608
:
VWR Collection
VWR®, Shoe Cover Dispenser
Catalog # 113-0608
Supplier:
VWR Collection
Local Article Number: VWRI113-0608
Discontinued
Specifications
Dispenser type:
Bench top
PPE type:
Shoe covers
Colour:
Silver
Description:
Shoe cover dispenser
W×D×H:
280×180×520 mm
Pk:
1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation