Northern Ireland
Order Entry
Supelco® Organic Reference Standard, Palmitoyl-ʟ-carnitine
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Palmitoyl-ʟ-carnitine
 :  Merck

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • CAS No.:
    2364-67-2
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt; ʟ-Carnitine hexadecanoyl ester; C16-Carnitine; Hexadecanoyl-ʟ-carnitine
  • Molecular weight:
    399,61

 

Product Information

  • Catalog #
    Supplier #
    Standards
    Pk
    Availability
    Price
  • Palmitoyl-ʟ-carnitine
    Palmitoyl-ʟ-carnitine
    91503-10MG
    91503-10MG
    Analytical standard
    10 mg
       
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £130.00
    £130.00
    1 * 10 mg
       £130.00

     : 91503-10MG
     : SUPL91503-10MG
  • Palmitoyl-ʟ-carnitine
    Palmitoyl-ʟ-carnitine
    91503-50MG
    91503-50MG
    Analytical standard
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - £414.00
    £414.00
    1 * 50 mg
       £414.00

     : 91503-50MG
     : SUPL91503-50MG