Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 555)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- AATB10380091
- AATB10380090
- Antigen symbol:
- CD38
- CD38
- Antigen name:
- CD38 antigen
- CD38 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 555
- iFluor® 555
- Clone:
- HIT2
- HIT2
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
- Pack typePk
- AATB1038009010380090Vial100 Tests1 * 100 Tests£328.00: 10380090: AATB10380090
Specifications
Antigen symbolCD38Antigen nameCD38 antigenEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationiFluor® 555CloneHIT2ReactivityHumanHostMouseIsotypeIgG1ImmunoFluorescenceYesFlow cytometryYesPurificationAffinity chromatographyCross adsorptionNoStorage temperature2...8 °C with minimized light exposure. Do not freeze.Shipping temperature20 - AATB1038009110380091Vial500 Tests1 * 500 Tests£1,230.00: 10380091: AATB10380091
Specifications
Antigen symbolCD38Antigen nameCD38 antigenEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationiFluor® 555CloneHIT2ReactivityHostMouseIsotypeIgG1ImmunoFluorescenceYesFlow cytometryYesPurificationAffinity chromatographyCross adsorptionNoStorage temperature2...8 °C with minimized light exposure. Do not freeze.Shipping temperature20