Highly cross-adsorbed donkey anti-rat IgG (H L) secondary antibody labeled with CF® Dyes.
- Highly cross-adsorbed for specific staining with minimal background
- Available in 13 bright and photostable CF® Dyes
- Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
This is a highly cross-adsorbed donkey anti-rat IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and sheep serum.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Donkey
Isotype:
Reactivity: Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 20896-500UL
- 20896-50UL
- Antigen name:
- Rat IgG
- Rat IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 583R
- CF® 583R
- Reactivity:
- Rat
- Rat
- Host:
- Donkey
- Donkey
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Liquid/lyophilised
- Liquid/lyophilised
- Concentration:
- 2 mg/ml
- 2 mg/ml
- Cross adsorption:
- Bovine, chicken, guinea pig, goat, horse, human, mouse, rabbit, sheep, Syrian hamster
- Bovine, chicken, guinea pig, goat, horse, human, mouse, rabbit, sheep, Syrian hamster
- Storage buffer:
- Liquid: PBS/50% glycerol/2 mg/ml BSA/0,5% azide
Lyophilised: PBS/15 mg/ml BSA/20 mg/ml trehalose after reconstitution - Liquid: PBS/50% glycerol/2 mg/ml BSA/0,5% azide
Lyophilised: PBS/15 mg/ml BSA/20 mg/ml trehalose after reconstitution
- Storage temperature:
- Store at –20 °C, protected from light
- Store at –20 °C, protected from light
- SeqSizePk
- New Product
Specifications
Antigen nameRat IgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationCF® 583RReactivityRatHostDonkeyIsotypeWestern blotYesImmunoFluorescenceYesFlow cytometryYesFormLiquid/lyophilisedConcentration2 mg/mlCross adsorptionBovine, chicken, guinea pig, goat, horse, human, mouse, rabbit, sheep, Syrian hamsterStorage bufferLiquid: PBS/50% glycerol/2 mg/ml BSA/0,5% azide
Lyophilised: PBS/15 mg/ml BSA/20 mg/ml trehalose after reconstitutionStorage temperatureStore at –20 °C, protected from lightNew Product - New Product
Specifications
Antigen nameRat IgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationCF® 583RReactivityRatHostDonkeyIsotypeWestern blotYesImmunoFluorescenceYesFlow cytometryYesFormLiquid/lyophilisedConcentration2 mg/mlCross adsorptionBovine, chicken, guinea pig, goat, horse, human, mouse, rabbit, sheep, Syrian hamsterStorage bufferLiquid: PBS/50% glycerol/2 mg/ml BSA/0,5% azide
Lyophilised: PBS/15 mg/ml BSA/20 mg/ml trehalose after reconstitutionStorage temperatureStore at –20 °C, protected from lightNew Product