CD8a (clone SK1) is a validated mouse monoclonal antibody that recognizes CD8a, a transmembrane glycoprotein and co-receptor found in T cells. This antibody derived from the CD8 alpha chain. The antibody is available conjugated to CF® Dyes and Astral™ tandem dyes. They are supplied in PBS, 0,1% BSA, 0,05% azide.
- Cell/tissue expression: Lymphocytes, T-cells
- Tumor expression: Leukemia/lymphoma
- SwissProt: P01732
- Unigene: 3003
- Positive control: Human peripheral blood mononuclear cells (PBMCs), CCRF-CEM or HL-60
- Ex/Em: 413/547 nm
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.
For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4 /CD8-. Anaplastic large cell lymphoma is usually CD4 and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation:CF®405L
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- P009-405L-125
- P009-405L-500
- Antigen symbol:
- CD8a
- CD8a
- Antigen synonyms:
- p32|Leu2|MAL|CD8
- p32|Leu2|MAL|CD8
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405L
- CF® 405L
- Clone:
- SK1
- SK1
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 925
- 925
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human peripheral blood T cells
- Human peripheral blood T cells
- Molecular weight:
- 32 - 34 kDa (alpha chain)
- 32 - 34 kDa (alpha chain)
- Storage buffer:
- PBS, 0,1% BSA, 0,05% azide
- PBS, 0,1% BSA, 0,05% azide
- Storage temperature:
- Store at 2...8 °C, protect fluorescent conjugates from light
- Store at 2...8 °C, protect fluorescent conjugates from light
- Shipping temperature:
- RT
- RT
- Tested applications:
- Flow (surface) (verified)
- Flow (surface) (verified)
- SizePk
- P009-405L-125P009-405L-125125 μl (25 tests)11 * 1 Each£153.00: P009-405L-125: BTIUP009-405L-125
Specifications
Antigen symbolCD8aEnvironmentally PreferableAntigen synonymsp32|Leu2|MAL|CD8Antibody typePrimaryClonalityMonoclonalConjugationCF® 405LCloneSK1ReactivityHostMouseGene ID925IsotypeIgG1 kappaFlow cytometryYesImmunogenHuman peripheral blood T cellsMolecular weight32 - 34 kDa (alpha chain)Storage bufferPBS, 0,1% BSA, 0,05% azideStorage temperatureStore at 2...8 °C, protect fluorescent conjugates from lightShipping temperatureRTTested applicationsFlow (surface) (verified) - P009-405L-500P009-405L-500500 μl (100 tests)11 * 1 Each£274.00: P009-405L-500: BTIUP009-405L-500
Specifications
Antigen symbolCD8aEnvironmentally PreferableAntigen synonymsp32|Leu2|MAL|CD8Antibody typePrimaryClonalityMonoclonalConjugationCF® 405LCloneSK1ReactivityHumanHostMouseGene ID925IsotypeIgG1 kappaFlow cytometryYesImmunogenHuman peripheral blood T cellsMolecular weight32 - 34 kDa (alpha chain)Storage bufferPBS, 0,1% BSA, 0,05% azideStorage temperatureStore at 2...8 °C, protect fluorescent conjugates from lightShipping temperatureRTTested applicationsFlow (surface) (verified)