Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Northern Ireland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
£0.00
Order Entry
Allophycocyanin fluorescent probe
Supplier:
AAT BIOQUEST
Certificates
Expand image
Allophycocyanin fluorescent probe
Supplier:
AAT BIOQUEST
About this item
:
:
105000 g/mol
:
5,II
:
GHS07
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
AATB2554
2554
Plastic bottle
1 mg
Log in
for availability
1 * 1 mg
1 * 1 mg - £106.00
£106.00
-
+
1 * 1 mg
£106.00
-
+
AATB2555
2555
Plastic bottle
10 mg
1 * 10 mg
1 * 10 mg - £338.00
£338.00
-
+
1 * 10 mg
£338.00
-
+