Hypersil GOLD™ Peptide HPLC Columns
Achieve consistent retention of all peptides, including hydrophilic ones, and resolve deamidated species. Designed for stability and lot-to-lot consistency, this column meets the needs of biopharmaceutical product development and manufacturing control.
- Consistent, reliable results
- Optimised peptide retention
- Unparalleled sequence coverage
- Diameter: 2,1 mm
- For use with: LC/MS
Hypersil GOLD Peptide™ column performance is assured through dual quality confirmation, including a peptide-based QC test, ensures confidence in ultrapure silica, ligand bonding, and column packing for unparalleled sequence coverage and higher peak capacity.
Efficiently separate key post-translational modifications like deamidation, crucial for in-depth protein analysis. Good peak shape for peptides using either FA- or TFA-containing mobile phase.
- Catalog No:
- HYPE26002-152130
- HYPE26002-102130
- HYPE26002-052130
- Carbon load:
- 11%
- 11%
- 11%
- Endcapped:
- Yes
- Yes
- Yes
- Max. pressure:
- 1500 bar
- 1500 bar
- 1000 bar
- pH range:
- 2 - 11
- 2 - 11
- 2 - 11
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- USP code:
- L1
- L1
- L1
- Column FormatPhaseColumn hardwarePore sizeParticle SizeLength
- HYPE26002-05213026002-052130AnalyticalReversed phaseFused silica175 Å1,9 µm50 mm1 * 1 items
Specifications
Environmentally PreferableCarbon load11%EndcappedYesMax. pressure1000 barpH range2 - 11Surface area220 m²/gTemperature limit60 °CUSP codeL1 - HYPE26002-10213026002-102130AnalyticalReversed phaseFused silica175 Å1,9 µm100 mm1 * 1 items
Specifications
Environmentally PreferableCarbon load11%EndcappedYesMax. pressure1500 barpH range2 - 11Surface area220 m²/gTemperature limit60 °CUSP codeL1 - HYPE26002-15213026002-152130AnalyticalReversed phaseFused silica175 Å1,9 µm150 mm1 * 1 items
Specifications
Environmentally PreferableCarbon load11%EndcappedYesMax. pressure1500 barpH range2 - 11Surface area220 m²/gTemperature limit60 °CUSP codeL1