Northern Ireland
Order Entry
Anti-COL2A1 Rabbit Polyclonal Antibody
Anti-COL2A1 Rabbit Polyclonal Antibody
 
undefined
Anti-COL2A1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ROCK600-401-104-05
  • ROCK600-401-104-01
  • Antigen symbol:
  • COL2A1
  • COL2A1
  • Antigen name:
  • Collagen alpha-1(II) chain
  • Collagen alpha-1(II) chain
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Bovine
  • Human, Bovine
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 1280
  • 1280
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Collagen Type II from human knee cartilage and bovine nasal cartilage.
  • Collagen Type II from human knee cartilage and bovine nasal cartilage.
  • Format:
  • IgG
  • IgG
  • Form:
  • Liquid (sterile filtered)
  • Liquid (sterile filtered)
  • Concentration:
  • 1.03 mg/mL by UV absorbance at 280 nm
  • 1.03 mg/mL by UV absorbance at 280 nm
  • Storage buffer:
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage temperature:
  • 4°C
  • 4°C
  • Shipping temperature:
  • 20
  • 20
  • Tested applications:
  • Dot Blot
  • Dot Blot

 

 

  •  
     
    Pk
     
     
  • ROCK600-401-104-05
    600-401-104-05
    0,5 mg
       
       
    1 * 0,5 mg
    1 * 0,5 mg - £1,510.00
    £1,510.00
    1 * 0,5 mg
       £1,510.00

     
     
    Specifications
    Antigen symbol
    COL2A1
    Antigen name
    Collagen alpha-1(II) chain
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Bovine
    Host
    Rabbit
    Gene ID
    1280
    Isotype
    ImmunoChemistry
    Yes
    Immunogen
    Collagen Type II from human knee cartilage and bovine nasal cartilage.
    Format
    IgG
    Form
    Liquid (sterile filtered)
    Concentration
    1.03 mg/mL by UV absorbance at 280 nm
    Storage buffer
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
    Storage temperature
    4°C
    Shipping temperature
    20
    Tested applications
    Dot Blot
  • ROCK600-401-104-01
    600-401-104-01
    0,1 mg
       
       
    1 * 0,1 mg
    1 * 0,1 mg - £787.00
    £787.00
    1 * 0,1 mg
       £787.00

     
     
    Specifications
    Antigen symbol
    COL2A1
    Antigen name
    Collagen alpha-1(II) chain
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Bovine
    Host
    Rabbit
    Gene ID
    1280
    Isotype
    ImmunoChemistry
    Yes
    Immunogen
    Collagen Type II from human knee cartilage and bovine nasal cartilage.
    Format
    IgG
    Form
    Liquid (sterile filtered)
    Concentration
    1.03 mg/mL by UV absorbance at 280 nm
    Storage buffer
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
    Storage temperature
    4°C
    Shipping temperature
    20
    Tested applications
    Dot Blot