Northern Ireland
Order Entry
Estradiol Related Compounds, Reference Standards
 :  USP
 
Estradiol Related Compounds, Reference Standards
 :  USP

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • CAS No.:
    571-92-6
  • Synonym(s):
    1,3,5(10)-estratrien-3,17β-diol-6-one
  • Formula:
    C₁₈H₂₂O₃
  • Description:
    Estradiol Related Compound C

 

Product Information

  • Catalog #
    Supplier #
    Standards
    Pack type
    Pk
    Availability
    Price
  • Estradiol Related Compound B
    Estradiol Related Compound B
    1250020.
    1250020
    USP reference standard
    Vial
    10 mg
       
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £1,180.00
    £1,180.00
    1 * 10 mg
       £1,180.00

    Supplier Article Number: 1250020
    Local Article Number: USPH1250020
  • Estradiol Related Compound C
    Estradiol Related Compound C
    1250041.
    1250041
    USP reference standard
    Vial
    15 mg
       
    Log in for availability
    1 * 15 mg
    1 * 15 mg - £1,180.00
    £1,180.00
    1 * 15 mg
       £1,180.00

    Supplier Article Number: 1250041
    Local Article Number: USPH1250041