Northern Ireland
Order Entry
Cefdinir, Reference Standards
 :  USP
 
Cefdinir, Reference Standards
Supplier:  USP

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    91832-40-5
  • Description:
    Cefdinir
  • Synonym(s):
    2(R)-2-[(Z)-2-(2-aminothiazol-4-yl)-2-(hydroxyimino)acetamido]-2-[(2RS,5RS)-5-methyl-7-oxo-2,4,5,7-tetrahydro-1H-furo[3,4-d][1,3]thiazin-2-yl]acetic acid

Specifications

Product Information

  • Catalog #
    Supplier #
    Standards
    Pack type
    Pk
    Availability
    Price
  • Cefdinir
    Cefdinir
    1097614.
    1097614
    USP reference standard
    Vial
    300 mg
       
    Log in for availability
    1 * 300 mg
    1 * 300 mg - £368.00
    £368.00
    1 * 300 mg
       £368.00

  • Cefdinir Related Compound A
    Cefdinir Related Compound A
    1097625.
    1097625
    USP reference standard
    Vial
    10 mg
       
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £1,210.00
    £1,210.00
    1 * 10 mg
       £1,210.00

  • Cefdinir Related Compound B
    Cefdinir Related Compound B
    1097589.
    1097589
    USP reference standard
    Vial
    10 mg
       
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £1,180.00
    £1,180.00
    1 * 10 mg
       £1,180.00