Northern Ireland
Order Entry
 
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), Pierce™
  PIER22589
 :  
undefined
 
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), Pierce™
  PIER22589
 :  
 : 22589
 : PIER22589
 
£497.00
   
  1 * 50 mg

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Sulfo-NHS Ester, Aryl Azide
  • Spacer:
    Long
  • Form:
    Solid
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

 

 :
 :492.40 g/mol
 :GHS07
 :Freezer
 :102568-43-4
 :C₁₆H₁₇O₉N₆NaS