Northern Ireland
Order Entry
 
Genipin
 
 
Genipin

 

  • Appearance:
    White to almost white powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    118.0°C to 123.0°C
  • HPLC:
    ≥ 98.0%
  • Water:
    ≤ 1.0%

 

 

  •  
     
    Pack type
    Pk
     
     
  • ACRO466642500
    466642500
    Glass bottle
    250 mg
       
       
    1 * 250 mg
    1 * 250 mg - £145.00
    £145.00
    1 * 250 mg
       £145.00

  • ACRO466640010
    466640010
    Glass bottle
    1 g
       
       
    1 * 1 g
    1 * 1 g - £407.00
    £407.00
    1 * 1 g
       £407.00