4-Bromo-3,5-difluoroaniline 97%
Supplier: Apollo Scientific
|
Warning
|
APOSPC0968-5GEA
14.3
GBP
APOSPC0968-5G
APOSPC0968-25G
4-Bromo-3,5-difluoroaniline 97%
4-Bromo-3,5-difluoroaniline
Formula:
C₆H₄BrF₂N MW: 208,01 g/mol Melting Pt: 99…101 °C Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236593 CAS Number: 203302-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...