1,14-Diamino-3,6,9,12-tetraoxatetradecane

Supplier: Apollo Scientific
Warning

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diamineTriethylene Glycol bis(2-aminoethyl) ether

APOSBIPG1056-250MGEA 321 GBP
APOSBIPG1056-250MG
1,14-Diamino-3,6,9,12-tetraoxatetradecane
1,14-Diamino-3,6,9,12-tetraoxatetradecane
Formula: C₁₀H₂₄N₂O₄
MW: 236,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01365938
CAS Number: 68960-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR