Latex gloves, Vigor®
Supplier: Sri Trang
112-6140EA 23.4 GBP
112-6140 112-6138 112-6139
Latex gloves, Vigor®
Gloves
These ambidextrous gloves are ideal for medical and lab applications, cleaning, and other tasks with low risks.
  • Non sterile
  • Powder-free
  • Textured fingertips for excellent grip
  • Rolled cuffs minimise the risk of tearing
  • Smooth and easy donning thanks to the inner polymer coating treatment
  • Level 2 – AQL 1,5
These high-quality latex gloves offer superior wearing comfort without leaving any residue. The glove exterior features an offline chlorination treatment.
Certifications: EN 455, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN 374-5; Cat. III; AQL 1,5
Food contact approved.