Anti-NEUN/FOX3 Rabbit Polyclonal Antibody
Supplier: Biosensis
BSENR-3771-100EA 0 GBP
BSENR-3771-100
Anti-NEUN/FOX3 Rabbit Polyclonal Antibody
Antibodies
Fox3 is one of a family of mammalian homologues of Fox-1. The Fox proteins are about 46 kDa in size, and each includes a central highly conserved RRM type RNA recognition motif. Much interest has focused on Fox3 as a result of the recent finding that this protein corresponds to NeuN, a neuronal nuclear antigen. NeuN/Fox-3 has a function in RNA splicing and is expressed heavily and specifically in neuronal nuclei and cytoplasm.
Applications: Western Blotting (WB), Immunocytochemistry (ICC) and Immunohistochemistry (IHC). A dilution of 1:500 - 1:1,000 is recommended for WB. A dilution of 1:5,000 - 1:10,000 is recommended for ICC and IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.