You Searched For: Spillage Trays/Pans


21  results were found

SearchResultCount:"21"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Avantor
Description: Spillage

Supplier: MEMMERT
Description: May affect the temperature distribution in the chamber.

Supplier: Bohlender
Description: Practical collecting tray made of PE or static dissipative PE-EX. Place your waste canister in a collecting tray. Escaping liquids are collected in ca...

Supplier: S.C.A.T.
Description: Made of HDPE, white or black (electrically conductive). Containers for all purposes.

Catalog Number: (NEWPPAN404)
Supplier: NEW PIG
Description: Light-weight, economical Utility Pans serve dozens of purposes throughout your workplace.
UOM: 1 * 12 items


Supplier: NEW PIG
Description: HDPE trays contain messy drips and spills from 210 litre drums.

Supplier: NEW PIG
Description: Catch drips and control messes with these smooth, seamless fibreglass trays.

Supplier: MEMMERT
Description: May affect the temperature distribution within the chamber.

Catalog Number: (NEWPPAK922)
Supplier: NEW PIG
Description: This durable, one-piece utility trays contain liquid messes from oily parts and leaky containers.
UOM: 1 * 1 items


Supplier: NEW PIG
Description: If containers leak, drip or crack, make sure a Mini Drip Deck with built-in sump is in position to catch the liquid.

Supplier: CEMO GMBH
Description: These small containers and collection trays can be used to carry and store water-polluting substances such as cleaning agents, pesticides, used oil fi...

Supplier: USBECK Laborgeräte
Description: Made of stainless steel.

Catalog Number: (NEWPPAKE272)
Supplier: NEW PIG
Description: Lightweight and durable PE spill trays are ideal for storing several small to medium sized containers of liquids.
UOM: 1 * 1 items


Supplier: JUSTRITE MANUFACTURING CO
Description: Safe and reliable for tabletop parts cleaning and fluid transfer operations involving petroleum products and acids, the Mini-Berm Flex Tray is made fr...

Catalog Number: (BELAF246760003)
Supplier: Bel-Art Products, a Part of SP
Description: This large, deep, polypropylene plastic tray protects users by capturing accidental hazardous spills.
UOM: 1 * 1 items


Supplier: JUSTRITE MANUFACTURING CO
Description: Eco-friendly spill trays are made from 100% post-industrial recycled PE will not rust or corrode.

Sale

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 21
no targeter for Bottom