You Searched For: Safety Shields


7  results were found

SearchResultCount:"7"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Bel-Art Products, a Part of SP
Description: The stable transparent safety shield PC (thickness: 4,7 mm) is constructed of clear PC barrier and heavy steel base is coated with blue epoxy coating.

Catalog Number: (113-2500)
Supplier: Bel-Art Products, a Part of SP
Description: These splash shields are designed to provide safety, comfort, flexibility and quality in a design that will adapt to your specific needs.
UOM: 1 * 1 items


Catalog Number: (113-2801)
Supplier: Thermo Fisher Scientific
Description: PC/acetate with magnetic base, transparent, thickness: 3 mm. Safety splash shield with flexible arm attaches easily to benchtops, shelving, machinery ...
UOM: 1 * 1 items


Supplier: KOEHLER TECHNISCHE PRODUKTEN
Description: Protective shield made from acrylic glass to be used as splinter shield and splash guard.

Supplier: TRIPP NT
Description: These draft shields have cube design with ball cap for deflecting downward drafts.

Catalog Number: (KART571)
Supplier: KARTELL
Description: PMMA protection shield, transparent, thickness: 5 mm, provides laboratory and healthcare personnel with facial protection from hazardous substances, a...
UOM: 1 * 1 items


Catalog Number: (ROTH2968.1)
Supplier: Roth Carl
Description: These safety shields are made from transparent acrylic glass of 8 mm thick.
UOM: 1 * 1 items

Market Source Item This is a MarketSource item. Additional charges may apply

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 7 of 7
no targeter for Bottom