Lithium chloride 2M in ethanol acc. ASTM D664 / IP 177 electrolytic solution
|
Danger
|
99166.001EA
81.8
GBP
99166.001
99167.001
99168.001
Lithium chloride 2M in ethanol acc. ASTM D664 / IP 177 electrolytic solution
Lithium chloride
A high quality analytical reagent manufactured and certified compliant for use in ASTM D664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
- A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
- Provided with a certificate which includes an estimation of uncertainty of manufacture.
- Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
- Convenient 30mL bottle size for direct filling into the electrode.
- Multi pack option available
- Cost effective and time saving
- Certificate of Manufacture & MSDS included
- 6 Month shelf life
A high quality analytical reagent manufactured and certified compliant for use in ASTM D664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
Formula:
LiCl MW: 42,39 g/mol Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...