Phytochemical standard, carnosic acid, analytical standard, Supelco®
Supplier: Merck
91209-5MGEA 198 GBP
91209-5MG
Phytochemical standard, carnosic acid, analytical standard, Supelco®
Standards Organic Standards
Carnosic acid is a polyphenolic diterpene, which can be obtained from rosemary. It may possess various pharmacological properties like antioxidant, anticancer and also antimutagenic property.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.