Light meter features a wide band wavelength of 320 to 390 nm.
- Traceable®
- Wide-band wavelength
- Measures on three different ranges
Can recall highest, lowest, and average readings.
Data acquisition system captures, displays, and stores readings on any PC. Information can be imported into databases. Includes 393,7 mm diskette and 1016 mm serial cable with D9F plug.
A 'Data Hold' button freezes the display to capture readings. Serial RS232 output allows meter to be connected to a computer or data logger for monitoring and storing results.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Light meter is supplied with UV sensor, 1016 mm cable, serial computer output, and 9 V battery.