/
Northern Ireland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Pharmaceutical secondary standard, Famciclovir, (CRM), Supelco®
PHR1696-1G
:
Merck
:
Pharmaceutical secondary standard, Famciclovir, (CRM), Supelco®
PHR1696-1G
:
Merck
:
£231.00
1 * 1 g
-
+
Pk:
1 g
CAS No.:
104227-87-4
Description:
Famciclovir
Environmentally Preferable:
Description:
Famciclovir