Required for brain and eye development. Promotes the disassembly of phosphorylated vimentin intermediate filaments (IF) during mitosis and may play a role in the trafficking and distribution of IF proteins and other cellular factors to daughter cells during progenitor cell division. Required for survival, renewal and mitogen-stimulated proliferation of neural progenitor cells (By similarity). Ref: uniprot.org
Western blotting (1:1,000-1:5,000) and Immunocytochemistry (1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NEST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human